Assay Details
| Gene Symbol: MED19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405407.1 | XP_022261115.1 | 1 - 2 | 445 | 64 | |
| XM_847563.5 | XP_852656.2 | 1 - 2 | 438 | 64 | ||
More Information
Set Membership: |
|

MED19
Genomic Map