Assay Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632868.2 | XP_005632925.1 | 4 - 5 | 482 | 74 | |
| XM_022407040.1 | XP_022262748.1 | 7 - 8 | 861 | 74 | ||
| XM_022407041.1 | XP_022262749.1 | 6 - 7 | 737 | 74 | ||
| XM_022407042.1 | XP_022262750.1 | 4 - 5 | 471 | 74 | ||
| XM_848818.4 | XP_853911.1 | 4 - 5 | 640 | 74 | ||
More Information
Set Membership: |
|

SPC24
Genomic Map