Assay Details
| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637176.2 | XP_005637233.1 | 4 - 5 | 618 | 73 | |
| XM_014108512.2 | XP_013963987.1 | 4 - 5 | 956 | 73 | ||
| XM_022411316.1 | XP_022267024.1 | 4 - 5 | 294 | 73 | ||
| XM_022411317.1 | XP_022267025.1 | 4 - 5 | 1067 | 73 | ||
More Information
Set Membership: |
|

KLRG1
Genomic Map