Assay Details
| Gene Symbol: LOC611915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621847.3 | XP_005621904.1 | 8 - 9 | 1194 | 91 | |
| XM_022420402.1 | XP_022276110.1 | 7 - 8 | 1086 | 91 | ||
| XM_022420403.1 | XP_022276111.1 | 8 - 9 | 1194 | 91 | ||
| XM_849638.5 | XP_854731.1 | 8 - 9 | 1125 | 91 | ||
More Information
Set Membership: |
|

LOC611915
Genomic Map