Assay Details
| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403883.1 | XP_022259591.1 | 1043 | 75 | ||
| XM_022403884.1 | XP_022259592.1 | 1046 | 75 | |||
More Information
Set Membership: |
|

GALNTL5
Genomic Map