Assay Details
| Gene Symbol: PGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116063.2 | XP_013971538.1 | 5 - 6 | 640 | 59 | |
| XM_014116065.2 | XP_013971540.1 | 4 - 5 | 601 | 59 | ||
| XM_014116067.2 | XP_013971542.1 | 6 - 7 | 773 | 59 | ||
| XM_014116071.2 | XP_013971546.1 | 2 - 3 | 460 | 59 | ||
| XM_022422220.1 | XP_022277928.1 | 7 - 8 | 1025 | 59 | ||
| XM_022422221.1 | XP_022277929.1 | 7 - 8 | 999 | 59 | ||
| XM_022422222.1 | XP_022277930.1 | 7 - 8 | 997 | 59 | ||
| XM_022422223.1 | XP_022277931.1 | 7 - 8 | 999 | 59 | ||
| XM_022422224.1 | XP_022277932.1 | 5 - 6 | 735 | 59 | ||
| XM_022422225.1 | XP_022277933.1 | 7 - 8 | 1025 | 59 | ||
| XM_022422226.1 | XP_022277934.1 | 7 - 8 | 989 | 59 | ||
| XM_022422227.1 | XP_022277935.1 | 7 - 8 | 986 | 59 | ||
| XM_022422228.1 | XP_022277936.1 | 2 - 3 | 462 | 59 | ||
More Information
Set Membership: |
|

PGF
Genomic Map