Assay Details
| Gene Symbol: ITFG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637311.3 | XP_005637368.1 | 5 - 6 | 918 | 66 | |
| XM_014108521.2 | XP_013963996.1 | 5 - 6 | 683 | 66 | ||
| XM_014108522.2 | XP_013963997.1 | 5 - 6 | 562 | 66 | ||
More Information
Set Membership: |
|

ITFG2
Genomic Map