Assay Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287081.1 | NP_001274010.1 | 2 - 3 | 112 | 89 | |
| GenBank mRNA | DN373580.1 | - | 2 - 3 | 112 | 89 | |
More Information
Set Membership: |
|

LSM7
Genomic Map