Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635104.3 | XP_005635161.1 | 2 - 3 | 456 | 141 | |
| XM_848514.5 | XP_853607.1 | 2 - 3 | 433 | 141 | ||
More Information
Set Membership: |
|

CCN5
Genomic Map