Assay Details
| Gene Symbol: STX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_849451.3 | XP_854544.1 | 340 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_849451.3 transcripts.
Set Membership: |
|

STX11
Genomic Map