Assay Details
| Gene Symbol: TP53I13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624889.3 | XP_005624946.1 | 6 - 7 | 1061 | 59 | |
| XM_005624890.3 | XP_005624947.1 | 6 - 7 | 1055 | 59 | ||
| XM_022423396.1 | XP_022279104.1 | 6 - 7 | 1020 | 59 | ||
| XM_022423397.1 | XP_022279105.1 | 4 - 5 | 591 | 59 | ||
| XM_022423398.1 | XP_022279106.1 | 3 - 4 | 381 | 59 | ||
| XM_022423399.1 | XP_022279107.1 | 4 - 5 | 419 | 59 | ||
| XM_022423400.1 | XP_022279108.1 | 4 - 5 | 418 | 59 | ||
| XM_022423401.1 | XP_022279109.1 | 5 - 6 | 631 | 59 | ||
| XM_022423402.1 | XP_022279110.1 | 4 - 5 | 529 | 59 | ||
| XM_022423403.1 | XP_022279111.1 | 5 - 6 | 650 | 59 | ||
| XM_022423404.1 | XP_022279112.1 | 2 - 3 | 318 | 59 | ||
| XM_848643.4 | XP_853736.1 | 4 - 5 | 420 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022423401.1, XM_022423402.1, XM_022423403.1, XM_022423404.1 transcripts.
Set Membership: |
|

TP53I13
Genomic Map