Assay Details
| Gene Symbol: C6H1orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_850261.5 | XP_855354.2 | 2 - 3 | 501 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6H1orf52| Gene Symbol: C6H1orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_850261.5 | XP_855354.2 | 2 - 3 | 501 | 111 | |
Set Membership: |
|