Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623674.3 | XP_005623731.1 | 3 - 4 | 438 | 65 | |
| XM_005623676.3 | XP_005623733.1 | 3 - 4 | 392 | 65 | ||
| XM_022422231.1 | XP_022277939.1 | 3 - 4 | 354 | 65 | ||
More Information
Set Membership: |
|

JDP2
Genomic Map