Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633135.3 | XP_005633192.1 | 8 - 9 | 1402 | 116 | |
| XM_014122295.2 | XP_013977770.1 | 7 - 8 | 1399 | 116 | ||
| XM_022407297.1 | XP_022263005.1 | 7 - 8 | 1407 | 116 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map