Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633135.3 | XP_005633192.1 | 11 - 12 | 1591 | 82 | |
| XM_014122295.2 | XP_013977770.1 | 10 - 11 | 1588 | 82 | ||
| XM_022407297.1 | XP_022263005.1 | 10 - 11 | 1596 | 82 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map