Assay Details
| Gene Symbol: BCAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107156.2 | XP_013962631.1 | 4 - 5 | 1096 | 84 | |
| XM_014107157.2 | XP_013962632.1 | 4 - 5 | 844 | 84 | ||
| XM_022408920.1 | XP_022264628.1 | 4 - 5 | 847 | 84 | ||
| XM_022408921.1 | XP_022264629.1 | 4 - 5 | 856 | 84 | ||
| XM_022408922.1 | XP_022264630.1 | 4 - 5 | 746 | 84 | ||
| XM_022408923.1 | XP_022264631.1 | 3 - 4 | 729 | 84 | ||
| XM_022408924.1 | XP_022264632.1 | 4 - 5 | 747 | 84 | ||
| XM_022408925.1 | XP_022264633.1 | 4 - 5 | 848 | 84 | ||
More Information
Set Membership: |
|

BCAS1
Genomic Map