Assay Details
| Gene Symbol: BTBD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119717.2 | XP_013975192.1 | 7 - 8 | 1337 | 66 | |
| XM_022428026.1 | XP_022283734.1 | 7 - 8 | 1335 | 66 | ||
| XM_022428027.1 | XP_022283735.1 | 7 - 8 | 770 | 66 | ||
More Information
Set Membership: |
|

BTBD19
Genomic Map