Assay Details
| Gene Symbol: SERPINF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077588.2 | NP_001071056.2 | 6 - 7 | 881 | 65 | |
| XM_005624362.3 | XP_005624419.1 | 6 - 7 | 942 | 65 | ||
| GenBank mRNA | DN411459.1 | - | 5 - 6 | 746 | 65 | |
More Information
Set Membership: |
|

SERPINF1
Genomic Map