Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623733.3 | XP_005623790.1 | 5 - 6 | 967 | 66 | |
| XM_005623734.3 | XP_005623791.1 | 4 - 5 | 889 | 66 | ||
| XM_005623735.3 | XP_005623792.1 | 4 - 5 | 779 | 66 | ||
| XM_005623736.3 | XP_005623793.1 | 4 - 5 | 948 | 66 | ||
| XM_005623737.3 | XP_005623794.1 | 5 - 6 | 967 | 66 | ||
| XM_022422311.1 | XP_022278019.1 | 5 - 6 | 897 | 66 | ||
| XM_022422312.1 | XP_022278020.1 | 4 - 5 | 879 | 66 | ||
| XM_022422314.1 | XP_022278022.1 | 4 - 5 | 777 | 66 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map