Assay Details
| Gene Symbol: LOC611813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405127.1 | XP_022260835.1 | 5 - 6 | 624 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC611813| Gene Symbol: LOC611813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405127.1 | XP_022260835.1 | 5 - 6 | 624 | 63 | |
Set Membership: |
|