Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619272.3 | XP_005619329.1 | 5 - 6 | 506 | 67 | |
| XM_014113053.2 | XP_013968528.1 | 5 - 6 | 551 | 67 | ||
More Information
Set Membership: |
|

SPARC
Genomic Map