Assay Details
| Gene Symbol: NMNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115125.2 | XP_013970600.1 | 1073 | 86 | ||
| XM_014115126.2 | XP_013970601.1 | 1307 | 86 | |||
| XM_022420960.1 | XP_022276668.1 | 1344 | 86 | |||
| XM_022420961.1 | XP_022276669.1 | 1408 | 86 | |||
| XM_022420962.1 | XP_022276670.1 | 901 | 86 | |||
| XM_022420963.1 | XP_022276671.1 | 1135 | 86 | |||
More Information
Set Membership: |
|

NMNAT2
Genomic Map