Assay Details
| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616336.3 | XP_005616393.1 | 6 - 7 | 1120 | 65 | |
| XM_022424188.1 | XP_022279896.1 | 6 - 7 | 1121 | 65 | ||
| XM_022424191.1 | XP_022279899.1 | 6 - 7 | 1122 | 65 | ||
More Information
Set Membership: |
|

IZUMO1
Genomic Map