Assay Details
| Gene Symbol: C24H20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409470.1 | XP_022265178.1 | 5 - 6 | 1283 | 74 | |
| XM_022409471.1 | XP_022265179.1 | 5 - 6 | 1225 | 74 | ||
| XM_022409472.1 | XP_022265180.1 | 6 - 7 | 1388 | 74 | ||
| XM_022409473.1 | XP_022265181.1 | 5 - 6 | 1188 | 74 | ||
| XM_022409474.1 | XP_022265182.1 | 4 - 5 | 1107 | 74 | ||
| XM_022409475.1 | XP_022265183.1 | 5 - 6 | 1948 | 74 | ||
More Information
Set Membership: |
|

C24H20orf173
Genomic Map