Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113113.2 | XP_013968588.1 | 2 - 3 | 1063 | 62 | |
| XM_014113114.2 | XP_013968589.1 | 2 - 3 | 1063 | 62 | ||
| XM_014113115.2 | XP_013968590.1 | 3 - 4 | 992 | 62 | ||
| XM_022417888.1 | XP_022273596.1 | 2 - 3 | 1063 | 62 | ||
More Information
Set Membership: |
|

PLCXD3
Genomic Map