Assay Details
| Gene Symbol: CFAP410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109834.2 | XP_013965309.1 | 6 - 7 | 844 | 141 | |
| XM_014109835.2 | XP_013965310.1 | 4 - 5 | 1769 | 141 | ||
| XM_022413213.1 | XP_022268921.1 | 7 - 8 | 937 | 141 | ||
| XM_849094.6 | XP_854187.5 | 6 - 7 | 737 | 141 | ||
More Information
Set Membership: |
|

CFAP410
Genomic Map