Assay Details
| Gene Symbol: RRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639270.3 | XP_005639327.1 | 6 - 7 | 3323 | 85 | |
| XM_022413490.1 | XP_022269198.1 | 6 - 7 | 3323 | 85 | ||
| XM_848046.4 | XP_853139.2 | 6 - 7 | 3322 | 85 | ||
More Information
Set Membership: |
|

RRH
Genomic Map