Assay Details
| Gene Symbol: ALDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107999.2 | XP_013963474.1 | 929 | 61 | ||
| XM_848535.5 | XP_853628.1 | 1 | 853 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ALDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107999.2 | XP_013963474.1 | 929 | 61 | ||
| XM_848535.5 | XP_853628.1 | 1 | 853 | 61 | ||
Set Membership: |
|