Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631439.3 | XP_005631496.1 | 2 - 3 | 336 | 62 | |
| XM_005631440.3 | XP_005631497.1 | 2 - 3 | 336 | 62 | ||
| XM_849643.5 | XP_854736.1 | 2 - 3 | 302 | 62 | ||
More Information
Set Membership: |
|

BRMS1
Genomic Map