Assay Details
| Gene Symbol: CA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145174.1 | NP_001138646.1 | 7 - 8 | 1068 | 57 | |
| XM_022424930.1 | XP_022280638.1 | 7 - 8 | 1110 | 57 | ||
More Information
Set Membership: |
|

CA9
Genomic Map