Assay Details
| Gene Symbol: TBC1D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640041.2 | XP_005640098.1 | 6 - 7 | 775 | 93 | |
| XM_005640043.3 | XP_005640100.1 | 5 - 6 | 692 | 93 | ||
| XM_005640045.2 | XP_005640102.1 | 4 - 5 | 620 | 93 | ||
| XM_022414471.1 | XP_022270179.1 | 5 - 6 | 737 | 93 | ||
| XM_535890.5 | XP_535890.1 | 5 - 6 | 700 | 93 | ||
More Information
Set Membership: |
|

TBC1D7
Genomic Map