Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640825.3 | XP_005640882.1 | 14 - 15 | 1741 | 109 | |
| XM_844375.5 | XP_849468.1 | 13 - 14 | 1719 | 109 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map