Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630689.3 | XP_005630746.1 | 19 - 20 | 2857 | 106 | |
| XM_005630690.3 | XP_005630747.1 | 12 - 13 | 3109 | 106 | ||
| XM_005630691.3 | XP_005630748.1 | 19 - 20 | 2860 | 106 | ||
| XM_005630692.3 | XP_005630749.1 | 17 - 18 | 2284 | 106 | ||
| XM_014120591.2 | XP_013976066.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120592.2 | XP_013976067.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120593.2 | XP_013976068.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120594.2 | XP_013976069.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120595.2 | XP_013976070.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120596.2 | XP_013976071.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120597.2 | XP_013976072.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120598.2 | XP_013976073.1 | 19 - 20 | 2860 | 106 | ||
| XM_014120599.2 | XP_013976074.1 | 19 - 20 | 2807 | 106 | ||
| XM_014120601.2 | XP_013976076.1 | 16 - 17 | 2479 | 106 | ||
| XM_014120602.2 | XP_013976077.1 | 19 - 20 | 2860 | 106 | ||
| XM_022404897.1 | XP_022260605.1 | 19 - 20 | 2860 | 106 | ||
| XM_022404898.1 | XP_022260606.1 | 12 - 13 | 3109 | 106 | ||
| XM_022404899.1 | XP_022260607.1 | 19 - 20 | 2860 | 106 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map