Assay Details
| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111037.2 | XP_013966512.1 | 5 - 6 | 503 | 72 | |
| XM_536018.6 | XP_536018.2 | 4 - 5 | 365 | 72 | ||
| XM_852308.5 | XP_857401.2 | 3 - 4 | 308 | 72 | ||
More Information
Set Membership: |
|

HSPE1
Genomic Map