Assay Details
| Gene Symbol: HPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635632.2 | XP_005635689.1 | 833 | 97 | ||
| XM_014107596.2 | XP_013963071.1 | 896 | 97 | |||
| XM_014107597.2 | XP_013963072.1 | 884 | 97 | |||
| XM_022409784.1 | XP_022265492.1 | 852 | 97 | |||
| XM_534550.6 | XP_534550.2 | 820 | 97 | |||
More Information
Set Membership: |
|

HPF1
Genomic Map