Assay Details
| Gene Symbol: LOC607405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619954.3 | XP_005620011.1 | 6 - 7 | 1569 | 74 | |
| XM_005619956.3 | XP_005620013.1 | 6 - 7 | 1525 | 74 | ||
| XM_022418478.1 | XP_022274186.1 | 5 - 6 | 782 | 74 | ||
| XM_022418479.1 | XP_022274187.1 | 5 - 6 | 1451 | 74 | ||
| XM_851891.5 | XP_856984.1 | 7 - 8 | 1642 | 74 | ||
More Information
Set Membership: |
|

LOC607405
Genomic Map