Assay Details
| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271782.1 | NP_001258711.1 | 3 - 4 | 370 | 53 | |
| XM_005641040.2 | XP_005641097.1 | 3 - 4 | 403 | 53 | ||
| XM_022415522.1 | XP_022271230.1 | 3 - 4 | 479 | 53 | ||
| XM_022415523.1 | XP_022271231.1 | 3 - 4 | 299 | 53 | ||
| XM_022415524.1 | XP_022271232.1 | 3 - 4 | 296 | 53 | ||
| GenBank mRNA | AB679834.1 | - | 4 - 5 | 594 | 53 | |
| DN372050.1 | - | 3 - 4 | 370 | 53 | ||
More Information
Set Membership: |
|

MPP1
Genomic Map