Assay Details
| Gene Symbol: C8H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623864.3 | XP_005623921.1 | 2 - 3 | 217 | 103 | |
| XM_005623865.3 | XP_005623922.1 | 2 - 3 | 217 | 103 | ||
| XM_005623866.3 | XP_005623923.1 | 2 - 3 | 234 | 103 | ||
| XM_005623867.3 | XP_005623924.1 | 2 - 3 | 276 | 103 | ||
| XM_005623868.3 | XP_005623925.1 | 2 - 3 | 256 | 103 | ||
| XM_005623869.3 | XP_005623926.1 | 2 - 3 | 217 | 103 | ||
| XM_005623870.3 | XP_005623927.1 | 3 - 4 | 207 | 103 | ||
| XM_005623871.3 | XP_005623928.1 | 3 - 4 | 347 | 103 | ||
| XM_014116172.2 | XP_013971647.1 | 2 - 3 | 263 | 103 | ||
| XM_014116174.2 | XP_013971649.1 | 2 - 3 | 1082 | 103 | ||
| XM_022422418.1 | XP_022278126.1 | 3 - 4 | 1118 | 103 | ||
| XM_022422419.1 | XP_022278127.1 | 2 - 3 | 217 | 103 | ||
| XM_022422420.1 | XP_022278128.1 | 2 - 3 | 217 | 103 | ||
More Information
Set Membership: |
|

C8H14orf180
Genomic Map