Assay Details
| Gene Symbol: TOB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625828.3 | XP_005625885.1 | 1 - 2 | 445 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005625828.3 transcripts.
Set Membership: |
|

TOB2
Genomic Map