Assay Details
| Gene Symbol: RARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636777.3 | XP_005636834.1 | 8 - 9 | 1286 | 72 | |
| XM_005636780.3 | XP_005636837.1 | 6 - 7 | 1327 | 72 | ||
| XM_014108423.2 | XP_013963898.1 | 6 - 7 | 973 | 72 | ||
| XM_022411224.1 | XP_022266932.1 | 6 - 7 | 987 | 72 | ||
| XM_853188.5 | XP_858281.4 | 6 - 7 | 1084 | 72 | ||
More Information
Set Membership: |
|

RARG
Genomic Map