Assay Details
| Gene Symbol: DLA-79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020810.1 | NP_001018646.1 | 3 - 4 | 635 | 71 | |
| XM_005631048.3 | XP_005631105.1 | 3 - 4 | 819 | 71 | ||
| XM_005631050.3 | XP_005631107.1 | 3 - 4 | 819 | 71 | ||
| XM_022405069.1 | XP_022260777.1 | 3 - 4 | 819 | 71 | ||
| XM_022405070.1 | XP_022260778.1 | 3 - 4 | 819 | 71 | ||
More Information
Set Membership: |
|

DLA-79
Genomic Map