Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625157.3 | XP_005625214.1 | 2 - 3 | 526 | 116 | |
| XM_014116959.2 | XP_013972434.1 | 2 - 3 | 547 | 116 | ||
| XM_852921.5 | XP_858014.1 | 2 - 3 | 472 | 116 | ||
More Information
Set Membership: |
|

BRD3
Genomic Map