Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625820.3 | XP_005625877.1 | 8 - 9 | 998 | 73 | |
| XM_014117312.2 | XP_013972787.1 | 9 - 10 | 2401 | 73 | ||
| XM_014117313.2 | XP_013972788.1 | 9 - 10 | 2403 | 73 | ||
| XM_022424352.1 | XP_022280060.1 | 10 - 11 | 2677 | 73 | ||
| XM_843889.5 | XP_848982.2 | 8 - 9 | 1012 | 73 | ||
More Information
Set Membership: |
|

SEPTIN3
Genomic Map