Assay Details
| Gene Symbol: TEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623901.3 | XP_005623958.1 | 4 - 5 | 621 | 61 | |
| XM_022422446.1 | XP_022278154.1 | 4 - 5 | 621 | 61 | ||
More Information
Set Membership: |
|

TEDC1
Genomic Map