Assay Details
| Gene Symbol: DARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406131.1 | XP_022261839.1 | 10 - 11 | 1178 | 72 | |
| XM_843573.5 | XP_848666.1 | 11 - 12 | 1237 | 72 | ||
More Information
Set Membership: |
|

DARS1
Genomic Map