Assay Details
| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628003.3 | XP_005628060.1 | 2 - 3 | 885 | 66 | |
| XM_005628004.3 | XP_005628061.1 | 2 - 3 | 1045 | 66 | ||
| XM_851288.6 | XP_856381.1 | 2 - 3 | 948 | 66 | ||
More Information
Set Membership: |
|

PTP4A3
Genomic Map