Assay Details
| Gene Symbol: SIRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631073.3 | XP_005631130.1 | 2 - 3 | 607 | 138 | |
| XM_005631077.3 | XP_005631134.1 | 2 - 3 | 607 | 138 | ||
| XM_014121034.2 | XP_013976509.1 | 2 - 3 | 2137 | 138 | ||
| XM_014121035.2 | XP_013976510.1 | 2 - 3 | 1969 | 138 | ||
| XM_014121037.2 | XP_013976512.1 | 2 - 3 | 1496 | 138 | ||
| XM_014121038.2 | XP_013976513.1 | 2 - 3 | 865 | 138 | ||
| XM_022405276.1 | XP_022260984.1 | 2 - 3 | 607 | 138 | ||
| XM_022405277.1 | XP_022260985.1 | 2 - 3 | 607 | 138 | ||
| XM_022405278.1 | XP_022260986.1 | 2 - 3 | 607 | 138 | ||
| XM_022405279.1 | XP_022260987.1 | 2 - 3 | 607 | 138 | ||
| XM_850716.5 | XP_855809.1 | 2 - 3 | 607 | 138 | ||
More Information
Set Membership: |
|

SIRT3
Genomic Map