Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633965.3 | XP_005634022.1 | 3 - 4 | 1140 | 95 | |
| XM_005633966.3 | XP_005634023.1 | 3 - 4 | 1077 | 95 | ||
| XM_843983.4 | XP_849076.1 | 3 - 4 | 1214 | 95 | ||
More Information
Set Membership: |
|

KLF5
Genomic Map