Assay Details
| Gene Symbol: ABHD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419553.1 | XP_022275261.1 | 667 | 65 | ||
| XM_546921.6 | XP_546921.1 | 787 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ABHD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419553.1 | XP_022275261.1 | 667 | 65 | ||
| XM_546921.6 | XP_546921.1 | 787 | 65 | |||
Set Membership: |
|