Assay Details
| Gene Symbol: ETV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616699.3 | XP_005616756.1 | 6 - 7 | 1468 | 69 | |
| XM_005616700.3 | XP_005616757.1 | 6 - 7 | 1288 | 69 | ||
| XM_005616702.3 | XP_005616759.1 | 5 - 6 | 1408 | 69 | ||
More Information
Set Membership: |
|

ETV2
Genomic Map